COM448634897 Combi-Blocks, Inc.

2-Propoxypyridine-3-boronic acid, pinacol ester

  • MDL number MFCD07781169
  • Cas Number : 1073371-87-5
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8894
Availability details for
Quantity
  • Availability
  • Add to Cart