COM448634899 Combi-Blocks, Inc.

5-(Morpholine-4-carbonyl)pyridine-3-boronic acid, pinacol ester

  • MDL number MFCD07781248
  • Cas Number : 1073371-92-2
  • Molecular Weight : 318.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8896
Availability details for
Quantity
  • Availability
  • Add to Cart