COM448634911 Combi-Blocks, Inc.

2-(4-Acetylpiperazin-1-yl)pyridine-5-boronic acid, pinacol ester

  • MDL number MFCD09027063
  • Cas Number : 1073372-01-6
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8910
Availability details for
Quantity
  • Availability
  • Add to Cart