COM448634933 Combi-Blocks, Inc.

2-(Dimethylaminocarbonyl)ethylboronic acid, pinacol ester

  • MDL number MFCD10700156
  • Cas Number : 134892-18-5
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8940
Availability details for
Quantity
  • Availability
  • Add to Cart