COM448634939 Combi-Blocks, Inc.

2-Chloro-5-hydroxymethylphenylboronic acid, pinacol ester

  • MDL number MFCD18729959
  • Cas Number : 1544673-40-6
  • Molecular Weight : 268.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9040
Availability details for
Quantity
  • Availability
  • Add to Cart