COM448634940 Combi-Blocks, Inc.

4-(Thiophen-2-yl)phenylboronic acid pinacol ester

  • MDL number MFCD27936848
  • Cas Number : 1056474-34-0
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9044
Availability details for
Quantity
  • Availability
  • Add to Cart