COM448634955 Combi-Blocks, Inc.

2-Morpholino-5-trifluoromethylphenylboronic acid, pinacol ester

  • MDL number MFCD09064984
  • Cas Number : 906352-77-0
  • Molecular Weight : 357.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9111
Availability details for
Quantity
  • Availability
  • Add to Cart