COM448634959 Combi-Blocks, Inc.

2-Acrylamidophenylboronic acid pinacol ester

  • MDL number MFCD10700144
  • Cas Number : 1218790-42-1
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9121
Availability details for
Quantity
  • Availability
  • Add to Cart