COM448634963 Combi-Blocks, Inc.

1-(P-Toluenesulfonyl)pyrrole-2-boronic acid pinacol ester

  • MDL number MFCD11045475
  • Cas Number : 1218790-43-2
  • Molecular Weight : 347.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9126
Availability details for
Quantity
  • Availability
  • Add to Cart