COM448634970 Combi-Blocks, Inc.

5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester

  • MDL number MFCD11113037
  • Cas Number : 1218790-45-4
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-9135
Availability details for
Quantity
  • Availability
  • Add to Cart