COM448634980 Combi-Blocks, Inc.

(2-Ethoxycarbonyl)phenylboronic acid, propanediol cyclic ester

  • MDL number MFCD06659932
  • Cas Number : 850567-60-1
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2126
Availability details for
Quantity
  • Availability
  • Add to Cart