COM448634981 Combi-Blocks, Inc.

2-(1,3,2-Dioxaborinan-2-yl)benzamide

  • MDL number MFCD20231486
  • Cas Number : 2377611-18-0
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2128
Availability details for
Quantity
  • Availability
  • Add to Cart