COM448634982 Combi-Blocks, Inc.

2-(3,4-Dimethylphenyl)-1,3,2-dioxaborinane, tech grade

  • MDL number MFCD09972182
  • Cas Number : 208390-39-0
  • Molecular Weight : 190.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2150
Availability details for
Quantity
  • Availability
  • Add to Cart