COM448634986 Combi-Blocks, Inc.

2-Methoxyphenylboronic acid, propanediol cyclic ester

  • MDL number MFCD06659934
  • Cas Number : 141522-26-1
  • Molecular Weight : 192.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2324
Availability details for
Quantity
  • Availability
  • Add to Cart