COM448634988 Combi-Blocks, Inc.

2-(Thiophen-2-yl)-1,3,2-dioxaborinane

  • MDL number MFCD11053858
  • Cas Number : 197024-83-2
  • Molecular Weight : 168.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2339
Availability details for
Quantity
  • Availability
  • Add to Cart