COM448634990 Combi-Blocks, Inc.

1-Phenylvinylboronic acid, propanediol cyclic ester

  • MDL number MFCD11504976
  • Cas Number : 415727-02-5
  • Molecular Weight : 188.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-2539
Availability details for
Quantity
  • Availability
  • Add to Cart