COM448634992 Combi-Blocks, Inc.

N-(2-Cyanoethyl)-4-(1,3,2-dioxaborinan-2-yl)benzamide

  • MDL number MFCD11053850
  • Cas Number : 1073372-08-3
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PR-3016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PR-3016
Availability details for
Quantity
  • Availability
  • Add to Cart