COM448634995 Combi-Blocks, Inc.

1H-Benzimidazole-2-carboxaldehyde, 5-methoxy-1-methyl-

  • MDL number MFCD11215482
  • Cas Number : 53004-19-6
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-0311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-0311
Availability details for
Quantity
  • Availability
  • Add to Cart