COM448635001 Combi-Blocks, Inc.

3-Bromo-2-piperazinopyridine

  • MDL number MFCD09910225
  • Cas Number : 87394-56-7
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1106
Availability details for
Quantity
  • Availability
  • Add to Cart