COM448635005 Combi-Blocks, Inc.

1-(5-Bromopyridin-3-yl)piperazine

  • MDL number MFCD09910230
  • Cas Number : 412347-30-9
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1113

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1113
Availability details for
Quantity
  • Availability
  • Add to Cart