COM448635006 Combi-Blocks, Inc.

1-(3-Bromo-5-methylpyridin-2-yl)piperazine

  • MDL number MFCD12546612
  • Cas Number : 1187386-35-1
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1115
Availability details for
Quantity
  • Availability
  • Add to Cart