COM448635014 Combi-Blocks, Inc.

2-Isobutoxypyridine

  • MDL number MFCD20278145
  • Cas Number : 1216841-32-5
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1125
Availability details for
Quantity
  • Availability
  • Add to Cart