COM448635021 Combi-Blocks, Inc.

2-(N-Benzyl-N-methylamino)-5-bromopyridine

  • MDL number MFCD12913988
  • Cas Number : 1187386-36-2
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1134
Availability details for
Quantity
  • Availability
  • Add to Cart