COM448635023 Combi-Blocks, Inc.

2-(4-Acetylpiperazino)-5-bromopyridine

  • MDL number MFCD11954763
  • Cas Number : 494771-76-5
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1136
Availability details for
Quantity
  • Availability
  • Add to Cart