COM448635032 Combi-Blocks, Inc.

Methyl 5-nitropicolinate

  • MDL number MFCD09062740
  • Cas Number : 29682-14-2
  • Molecular Weight : 182.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1148
Availability details for
Quantity
  • Availability
  • Add to Cart