COM448635036 Combi-Blocks, Inc.

5-bromo-2-(tert-butoxy)pyrimidine

  • MDL number MFCD09037471
  • Cas Number : 121487-13-6
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1152

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1152
Availability details for
Quantity
  • Availability
  • Add to Cart