COM448635038 Combi-Blocks, Inc.

5-Bromo-2-butoxypyrimidine

  • MDL number MFCD15143619
  • Cas Number : 1215206-57-7
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1154
Availability details for
Quantity
  • Availability
  • Add to Cart