COM448635048 Combi-Blocks, Inc.

2-(4-BOC-piperazino)-3-bromo-5-methylpyridine

  • MDL number MFCD12913989
  • Cas Number : 1187386-02-2
  • Molecular Weight : 356.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1165
Availability details for
Quantity
  • Availability
  • Add to Cart