COM448635053 Combi-Blocks, Inc.

5-Bromo-2-pivaloylamino-3-picoline

  • MDL number MFCD09037447
  • Cas Number : 446299-84-9
  • Molecular Weight : 271.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1174
Availability details for
Quantity
  • Availability
  • Add to Cart