COM448635064 Combi-Blocks, Inc.

5-Bromo-2-(Cbz-amino)pyridine

  • MDL number MFCD11976386
  • Cas Number : 175393-09-6
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1193
Availability details for
Quantity
  • Availability
  • Add to Cart