COM448635065 Combi-Blocks, Inc.

5-Bromo-2-(methoxycarbonylamino)pyridine

  • MDL number MFCD00739213
  • Cas Number : 207922-56-3
  • Molecular Weight : 231.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1194
Availability details for
Quantity
  • Availability
  • Add to Cart