COM448635087 Combi-Blocks, Inc.

5-Bromo-3-chloro-2-cyclohexylaminopyridine

  • MDL number MFCD18783192
  • Cas Number : 1280786-98-2
  • Molecular Weight : 289.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1224
Availability details for
Quantity
  • Availability
  • Add to Cart