COM448635094 Combi-Blocks, Inc.

3-Bromo-2-propoxypyridine

  • MDL number MFCD09743637
  • Cas Number : 760207-92-9
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1236
Availability details for
Quantity
  • Availability
  • Add to Cart