COM448635099 Combi-Blocks, Inc.

4-Bromo-2-mercapto-6-(trifluoromethoxy)benzothiazole

  • MDL number MFCD13195780
  • Cas Number : 1215206-47-5
  • Molecular Weight : 330.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1242
Availability details for
Quantity
  • Availability
  • Add to Cart