COM448635105 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-pyrrolidinopyridine

  • MDL number MFCD13195785
  • Cas Number : 1199773-44-8
  • Molecular Weight : 261.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1248
Availability details for
Quantity
  • Availability
  • Add to Cart