COM448635109 Combi-Blocks, Inc.

N-(4-Methyl-3-nitropyridin-2-yl)acetohydrazide

  • MDL number MFCD13195787
  • Cas Number : 1215205-33-6
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1252
Availability details for
Quantity
  • Availability
  • Add to Cart