COM448635121 Combi-Blocks, Inc.

2-Acetamido-5-bromo-3-nitropyridine

  • MDL number MFCD00770745
  • Cas Number : 381679-24-9
  • Molecular Weight : 260.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1266
Availability details for
Quantity
  • Availability
  • Add to Cart