COM448635124 Combi-Blocks, Inc.

2-(N'-Acetylhydrazino)-5-bromopyridine

  • MDL number MFCD13195789
  • Cas Number : 1199773-29-9
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1269
Availability details for
Quantity
  • Availability
  • Add to Cart