COM448635130 Combi-Blocks, Inc.

5-Bromo-2-(ethylthio)pyrimidine

  • MDL number MFCD13188492
  • Cas Number : 859941-10-9
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1278
Availability details for
Quantity
  • Availability
  • Add to Cart