COM448635143 Combi-Blocks, Inc.

N-tert-Butylisonicotinamide

  • MDL number MFCD00466497
  • Cas Number : 65321-30-4
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1295
Availability details for
Quantity
  • Availability
  • Add to Cart