COM448635145 Combi-Blocks, Inc.

N-tert-Butyl-2-chloroisonicotinamide

  • MDL number MFCD11128609
  • Cas Number : 588694-28-4
  • Molecular Weight : 212.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1297
Availability details for
Quantity
  • Availability
  • Add to Cart