COM448635154 Combi-Blocks, Inc.

2-Chloro-6,8-dimethylquinoline

  • MDL number MFCD15143624
  • Cas Number : 139719-23-6
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1306

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1306
Availability details for
Quantity
  • Availability
  • Add to Cart