COM448635157 Combi-Blocks, Inc.

8-Fluoroquinoline HCl

  • MDL number MFCD15143626
  • Cas Number : 311346-65-3
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1309
Availability details for
Quantity
  • Availability
  • Add to Cart