COM448635158 Combi-Blocks, Inc.

2-Ethoxyquinoline

  • MDL number MFCD00234738
  • Cas Number : 46185-83-5
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1310
Availability details for
Quantity
  • Availability
  • Add to Cart