COM448635171 Combi-Blocks, Inc.

6-Bromo-4-methyl-1,4-benzoxazin-3-one

  • MDL number MFCD08669851
  • Cas Number : 24036-47-3
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1326
Availability details for
Quantity
  • Availability
  • Add to Cart