COM448635174 Combi-Blocks, Inc.

2-Methoxy-6-phenylpyridine

  • MDL number MFCD11044374
  • Cas Number : 35070-08-7
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1329
Availability details for
Quantity
  • Availability
  • Add to Cart