COM448635175 Combi-Blocks, Inc.

6-Phenylpyridin-2(1H)-one

  • MDL number MFCD00059760
  • Cas Number : 19006-82-7
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1330
Availability details for
Quantity
  • Availability
  • Add to Cart