COM448635183 Combi-Blocks, Inc.

5-Bromo-(2-morpholinoethylamino)pyridine

  • MDL number MFCD09910232
  • Cas Number : 364794-56-9
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1344
Availability details for
Quantity
  • Availability
  • Add to Cart