COM448635187 Combi-Blocks, Inc.

2-Methoxy-6,8-dimethylquinoline

  • MDL number MFCD15143628
  • Cas Number : 861581-28-4
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1348
Availability details for
Quantity
  • Availability
  • Add to Cart