COM448635195 Combi-Blocks, Inc.

3-Bromo-5-fluoropyridine 1-oxide

  • MDL number MFCD16036155
  • Cas Number : 1221793-60-7
  • Molecular Weight : 192.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1358
Availability details for
Quantity
  • Availability
  • Add to Cart